LaMotte® Acidity Test Kit
LaMotte® Acidity Test Kit
Soil Acidity Test Kit, 4 - 8 pH
Catalog # 470201-278
Supplier:   LaMotte
Supplier #:  5024
LaMotte® Acidity Test Kit
Catalog # 470201-278
Supplier:  LaMotte
CAS Number:  
LaMotte® Acidity Test Kit
LaMotte® Acidity Test Kit
Soil Acidity Test Kit, 4 - 8 pH
CAS Number:  
Price
$52.35
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Accurate pH Testing
  • 100 Tests
  • Spot Testing of Your Soil
Tests are made with a wide range indicator and give the pH of soil to ±0.5 pH units. Includes 50 tests, chemical indicator, porcelain plates, color comparator chart, complete Instructions, pocket-size case and a soil test handbook.
Specifications
Specifications
  • Désignation:
    Soil Acidity Test Kit
  • Range:
    4 - 8 pH