Surgical Scissors
Surgical Scissors, 165 mm (6.5"), Stainless steel, Sharp/Blunt
Catalog # 470201-144
Supplier: Dunrite Instruments
Fournisseur #: WLS26975-C
Jump to:
Product Details & Documents
Each feature a sharp point for penetration and a blunt point for situations where not desired.
Available in four sizes for most dissection needs.
: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
- Désignation:Surgical Scissors
- Longueur:165 mm (6.5")
- Matière:Stainless steel
- Blade Type:Sharp/Blunt