Three-Oscilloscope Probe Kit
Three-Oscilloscope Probe Kit
470201-002
: CT3033
Designed for use with general purpose oscilloscopes.
This convenient and cost-effective kit contains three high-performance, passive voltage probes ideal for a wide variety of electronic, electrical, medical, and industrial applications.
Choose between a versatile 1×/10× switchable probe, a 1× fixed probe, or a high voltage 100× fixed attenuation probe. All feature low noise figures, durable construction, slim bodies, and unique snap-locking sprung hooks.
: Kit contents are packaged in a plastic carrying case for convenient transport and storage.
- Désignation:Three-Oscilloscope Probe Kit