Document Analysis Lab Activity
  470200-744
Document Analysis Lab Activity
  470200-744
 :  
Document Analysis Lab Activity
Document Analysis Lab Activity
REFILL KIT ANALYSIS OF DOC LAB ACT  470015-368
Product 470015-368 is a replacement for 470200-744 and has the same packaging and specifications, but may be a different quantity.
Determine a document’s authenticity.
  • Grades 6-12
  • Analysis of secret messages using chemistry
Your students will be fascinated as they perform both physical and chemical analysis of handwriting materials found at a crime scene. The procedures they will follow cover checking for erasures, abnormal positioning of a signature, use of different inks, and differences in handwriting or typing. It comes with enough materials for six setups and an instruction manual. A UV lamp, required, is available separately.