Complete Precision Air Table | Avantor
Complete Precision Air Table
Complete Precision Air Table
Catalog # 470200-444
Complete Precision Air Table
Catalog # 470200-444
Supplier:  
Complete Precision Air Table
Complete Precision Air Table
Complete Precision Air Table
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
AIR TABLE COMPLETE PRECISION (2 BOXES) Catalog #470006-994
Le produit 470006-994 est similaire à 470200-444 mais le fabricant et les spécifications peuvent varier.
AIR TABLE PRECISION Catalog #470157-648
Le produit 470157-648 est similaire à 470200-444 mais le fabricant et les spécifications peuvent varier.
Product Details & Documents
The Complete Precision Air Table helps investigate low-friction collisions.
  • Instructions with Suggested Experiments Included
  • Air Table Made of Heavy Glass
  • Air Compressor Included
  • Spark Sources Works with Six Frequencies
The recording system on this air table makes it easy to take good quantitative measurements from an air-table experiment.
A spark from the spark generator leaves a mark on the recording paper placed in the table. Students then use a protractor and centimeter scale to take measurements. The spark source can be operated at six different frequencies.
Storage recommendation for Carbon paper: Carbon papers should be stored in a dry place as the backing paper is sensitive to dampness. If not stored properly, the backing sheet will be difficult to remove from the carbon sheet.
Certifications: CSA approved.
Delivery information: This kit includes an air table made of heavy glass with a metal frame, air compressor, aluminum support for air hoses, air-supply hose, spark generator, two 550 g steel pucks, one 160 g auxiliary mass for unequal mass collisions, Velcro adhesive puck collars, two springs, four table pins, one orbital post with suction cup, two wire collars with hooks for pucks, a pulley, 100 sheets of recording paper, one sheet of carbon paper, and an instruction manual with suggested experiments.
Spécifications
  • Désignation:
    Complete Precision Air Table