Ward's® Student Dissecting Set
Student Dissecting Set Leatherette Case, 7 Instruments, 9, 1
Catalog # 470200-180
Supplier: Dunrite Instruments
Supplier #: WLS1407-71
Jump to:
Product Details & Documents
Great for high school dissection applications.
- Durable stainless steel tools
- Protective plastic case
We have conveniently collected all the instruments necessary for most high school dissection applications. Choose from a molded plastic or an improved, high-quality leatherette storage case for these fine instruments.
Packaging: Please see datasheet for contents.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
- Désignation:Student Dissecting Set Leatherette Case
- No. of Lab Groups:1
- Includes:7 Instruments
- No. of Parts:9