Document Analysis Lab Activity
Document Analysis Lab Activity
Catalog # 470200-098
Document Analysis Lab Activity
Catalog # 470200-098
Supplier:  
Document Analysis Lab Activity
Document Analysis Lab Activity
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
ANALYSIS OF DOCUMENTS LAB ACT. 24 STUD Catalog #470002-510
Product 470002-510 is a replacement for 470200-098 and has the same packaging and specifications, but may be a different quantity.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Determine a document’s authenticity.
  • Grades 6-12
  • Analysis of secret messages using chemistry
Your students will be fascinated as they perform both physical and chemical analysis of handwriting materials found at a crime scene. The procedures they will follow cover checking for erasures, abnormal positioning of a signature, use of different inks, and differences in handwriting or typing. It comes with enough materials for six setups and an instruction manual. A UV lamp, required, is available separately.