Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Aluminium Oxide 60 G Neutral (type E) for Thin-Layer Chromatography, Merck

Aluminium Oxide 60 G Neutral (type E) for Thin-Layer Chromatography, Merck

2.5 kg, Plastic Bottle
Catalog # CA1.01090.2500
Supplier:   MilliporeSigma
Supplier #:  1.01090.2500
Aluminium Oxide 60 G Neutral (type E) for Thin-Layer Chromatography, Merck
Catalog # CA1.01090.2500
Supplier:  MilliporeSigma
CAS Number:  
Aluminium Oxide 60 G Neutral (type E) for Thin-Layer Chromatography, Merck

2.5 kg, Plastic Bottle

CAS Number:  
Price
$3,814.11
Log in pour la disponibilité
Each (2,500g) Case of 4 (2,500,g)
Product Details & Documents

Standardized sorbents ensure reliable results every time

EMD Millipore's broad spectrum meets all kinds of needs. In TLC, silica gel 60 sorbent is considered the most versatile. Our assortment of silica gel 60 features different grades with particle sizes ranging from 5-40 µ: silica with a gypsum binder, silica with no foreign binder, and silica gel with a fluorescence indicator. We also offer high-quality aluminum oxide, cellulose microcrystalline, and kieselgur (also known as diatomaceous earth).

Spécifications

Spécifications

  • Type de conditionnement:
    Plastic Bottle
  • Taille:
    2.5 kg

Spécifications