Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VWR® Cleanroom Papers | Avantor

VWR® Cleanroom Papers

Latex-free paper, White, 8.5×14", 22 lbs.
Catalog # 89422-732
Supplier:   VWR International
VWR® Cleanroom Papers
Catalog # 89422-732
Supplier:  VWR International
VWR® Cleanroom Papers
VWR® Cleanroom Papers

Latex-free paper, White, 8.5×14", 22 lbs.

Price
$421.61
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 1,250
Product Details & Documents

The cleanroom paper is ideal for documentation in critical environments, where very low particle and ESD generation is important.

  • Low particle generation

Cleanroom paper is provided in latex or latex-free variants in a variety of colors.

Spécifications
  • Color:
    White
  • Désignation:
    Latex-free paper
  • Dimensions:
    8.5×14"
  • Weight:
    22 lbs.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.