Jars and Lids
Lid, 53 mm, for 2 oz. Jar #29488, Screw Type, Smooth, Unlined, Plastic, Black/White
Catalog # 470191-524
Supplier: PARKWAY PLASTICS, INC.
Supplier #: C053C4SPLIW
Jump to:
Product Details & Documents
Stock up on classroom essentials for every classroom activity.
Specifications
- Color:Black/White
- Désignation:Lid, 53 mm, for 2 oz. Jar #29488, Screw Type, Smooth, Unlined
- Matière:Plastic
Accessories
Product Information
- Catalog #AvailabilityPrice
- Screw Cap for Specimen Jar, 80 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



