Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Standard-Grade Glass Beaker | Avantor

Standard-Grade Glass Beaker

800 ml, 50 ml, 50 - 750 ml
Catalog # 470191-266
Supplier:   VWR International
Standard-Grade Glass Beaker
Catalog # 470191-266
Supplier:  VWR International
Standard-Grade Glass Beaker
Standard-Grade Glass Beaker

800 ml, 50 ml, 50 - 750 ml

Price
$12.00
Log in pour la disponibilité
Each Case of 24
Product Details & Documents

Our high-quality borosilicate beakers meet ASTM standards and are a great value. With uniform wall thickness, white enamel, double-graduated metric scale, and a marking spot. Beakers can be heated and used with acids, salt solutions, and organic solvents.

Documents

Spécifications
  • Intervalle de graduation:
    50 ml
  • Plage de graduation:
    50 - 750 ml
  • Capacité:
    800 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.