Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Standard-Grade Glass Beaker

30 ml, 10 ml, 5 - 25 ml
Catalog # 470191-146
Supplier:   VWR International
Standard-Grade Glass Beaker
Catalog # 470191-146
Supplier:  VWR International
Standard-Grade Glass Beaker
Standard-Grade Glass Beaker

30 ml, 10 ml, 5 - 25 ml

Price
$9.15
Log in pour la disponibilité
Each Case of 48
Product Details & Documents

Our high-quality borosilicate beakers meet ASTM standards and are a great value. With uniform wall thickness, white enamel, double-graduated metric scale, and a marking spot. Beakers can be heated and used with acids, salt solutions, and organic solvents.

Spécifications
  • Intervalle de graduation:
    10 ml
  • Plage de graduation:
    5 - 25 ml
  • Capacité:
    30 ml