Fast Release Pi-Pumps | Avantor
Fast Release Pi-Pumps
Fast Release Pi-Pump, 25 ml
Catalog # 470145-288
Fournisseur #:  F37904-0025
Fast Release Pi-Pumps
Catalog # 470145-288
Supplier:  
Fast Release Pi-Pumps
Fast Release Pi-Pumps
Fast Release Pi-Pump, 25 ml
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
PIPETTE PUMP ERGON 25 ML RED FAST REL Catalog #470153-802
Le produit 470153-802 est un remplacement de 470145-288 et a le même emballage et les mêmes spécifications, mais peut être d'une quantité différente.
Product Details & Documents
Designed for fast and accurate one-handed operation, this pump will reduce the risk of injury from repetitive pipetting.
  • Each pump is resistant to acids, bases, and most solvents
  • Disassembly for cleaning is easy
The fast-release trigger expels the entire contents at one shot, or you can use the thumb wheel for precision dispensing.
Spécifications
  • Capacité:
    25 ml
  • Désignation:
    Fast Release Pi-Pump