Funnel Sets
Funnel Set, Set of Three
Catalog # 470175-294
Supplier: Hutzler Manufacturing
Supplier #: 248
Jump to:
Product Details & Documents
Each set includes three funnels of different sizes. Small funnels are made of polypropylene; large funnels are made of polyethylene. Sizes in small funnel set: 60 mm top dia. x 20 mm stem, 80 mm top dia. x 22 mm stem, 110 mm top dia. x 36 mm stem. Sizes in large set: 127 mm top dia. x 45 mm stem, 160 mm top dia. x 50 mm stem, 200 mm top dia. x 60 mm stem.
Spécifications
- Désignation:Funnel Set, Set of Three