Ward's® Graphite | Avantor
Ward's® Graphite
Graphite (Pure), Bulk Pack, 1 kg, Size will vary
Catalog # 470025-596
Supplier:   Avantor
Ward's® Graphite
Catalog # 470025-596
Supplier:  Avantor
Ward's® Graphite
Ward's® Graphite
Graphite (Pure), Bulk Pack, 1 kg, Size will vary
Price
$96.35
Log in pour la disponibilité
Each
Product Details & Documents
Black, high grade, foliated, greasy feel.
  • Collected in Sri Lanka
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Spécifications
  • Désignation:
    Graphite (Pure), Bulk Pack, 1 kg
  • Specimen Dimensions:
    Size will vary