Plastic Vials
Vials, with caps, Oui
Catalog # 470020-892
Supplier: Avantor
Jump to:
Product Details & Documents
Made of safe, unbreakable plastic, the vials are available with or without friction-fit caps to prevent escape.
- 3 oz. capacity
Use our foam plugs for ventilation. Size: 4×1 ³/₈".
Caution: Note: Not suitable for holding liquid.
Spécifications
- Avec bouchon:Oui
- Désignation:Vials, with caps