Cellophane
Cellophane Sheets, Red, 20×60"
Catalog # 470020-514
Supplier: Avantor
Jump to:
Product Details & Documents
Use the various colors to demonstrate plants’ reaction to light waves.
- Made from recycled materials
Also useful for demonstrating the effects of mixing various colors of light.
Spécifications
- Color:Red
- Désignation:Cellophane Sheets
- Taille:20×60"