Plastic Beads
Plastic Beads, 4 mm
Catalog # 470015-258
Supplier: Avantor
Jump to:
Product Details & Documents
Packages of approximately 100.
- 4 mm Red bead = Acrylic
- 8 mm Navy bead = Acrylic
- 12 mm White bead = Acetate
- Autoclavable
Spécifications
- Ø:4 mm
- Désignation:Plastic Beads