Snap Circuits Educational Electronics Training Programs
Snap Circuits Educational Electronics Training Programs
Snap Circuits, SC100R Educational, Electronics Training for the Elementary Level
Catalog # 470105-896
Supplier:   Elenco Electronics
Supplier #:  SC-100R-A
Snap Circuits Educational Electronics Training Programs
Catalog # 470105-896
Supplier:  Elenco Electronics
Snap Circuits Educational Electronics Training Programs
Snap Circuits Educational Electronics Training Programs
Snap Circuits, SC100R Educational, Electronics Training for the Elementary Level
Price
$150.15
Log in for availability
Kit of 1
Product Details & Documents
The award-winning Snap Circuit Series uses building blocks with snaps to build electrical and electronic circuit projects.
  • Experiment Manual Included
  • Five Hundred Experiments Included
  • Storage Box Included
  • Experiment Topics Include Basic Components and Circuits, Capacitors, and More
  • Series Uses Building Blocks with Snaps to Build Electrical Projects
Each block has a function, and the blocks are color coded and numbered so they're easy to identify.
The blocks feature 500 experiments including basic components and circuits, motors and electricity, resistance, capacitors, transistors, oscillators and electronic sound, integrated circuits, electromagnetism, and radio. Other experiments cover meters, transformers, and FM radio; diodes and applications; and electronic switches.
Ordering information: This kit includes an experiment manual, student guide, and parts organizer/storage box. It requires two AA batteries (not included).
Specifications
  • Désignation:
    Snap Circuits, SC100R Educational, Electronics Training for the Elementary Level
  • Cat. No.:
    470105-896