Edvotek® Variable Micropipettes
20 - 200 µl, 0.1 µl
Catalog # 470103-994
Supplier: Edvotek
Supplier #: 591-1
Jump to:
Product Details & Documents
Our newly re-designed variable micropipettes are sturdily designed with volumes ranging from 0.1 to 5000 µl.
- New and improved
- High quality, same price
- More durable and ergonomic
- Lifetime warranty
- Light gray color
They are easy to use, highly accurate and use standard micropipette tips. The volume is easily selected by twisting the top. The lightweight design and tip ejector make operation fast and easy.
Ordering information: A tool and instructions are included for self-calibration.
Specifications
- Volume:20 - 200 µl
- Increments:0.1 µl