Boreal Standard Compound Microscopes
Compound microscope, Iris, LED cordless, Mechanical
Catalog # 470014-588
Supplier:   VWR
 :  MSH008
Boreal Standard Compound Microscopes
Catalog # 470014-588
Supplier:  VWR
Boreal Standard Compound Microscopes
Boreal Standard Compound Microscopes
Compound microscope, Iris, LED cordless, Mechanical
Price
$590.65
Log in pour la disponibilité
Each
Product Details & Documents
Boreal standard microscopes feature metal construction, a rugged student-proof design, and precision optics making them a favorite among educators.
  • Coated-glass optics
  • Wide-field eyepiece with pointer
  • Limited lifetime warranty
These microscopes optics are parfocal and parcentered for a crisp, consistent field of view. All units feature strategically located controls, course and fine focus, and a 360° rotatable head. Units are available in a variety of configurations offering three or four objectives, plain or mechanical stage, and corded or cordless charging.
Ordering information: Microscopes include a manual, dust cover, and limited warranty.
Spécifications
  • Désignation:
    Compound microscope
  • Stage:
    Mechanical
  • Condenser Type:
    Iris
  • Illumination:
    LED cordless
  • Cat. No.:
    470014-588
  • Objectives:
    4×, 10×, 40×
Accessories
Informations sur le produit
  • Catalog #
    Electrical
    Availability
    Price
  • Replacement bulb
    Replacement bulb
    470005-050
    1 W
    Log in pour la disponibilité
    Each - $19.45
Informations sur le produit
  • Catalog #
    Electrical
    Availability
    Price
  • Replacement Bulb, with Bayonet Base
    Replacement Bulb, with Bayonet Base
    470021-694
    110 V, 20 W
    Log in pour la disponibilité
    Each - $29.45