Ward's® Grassfrog Dissection Lab | Avantor
Ward's® Grassfrog Dissection Lab
WARD’S Grassfrog Dissection Lab
Catalog # 470027-814
Supplier:   Avantor
Ward's® Grassfrog Dissection Lab
Catalog # 470027-814
Supplier:  Avantor
Ward's® Grassfrog Dissection Lab
Ward's® Grassfrog Dissection Lab
WARD’S Grassfrog Dissection Lab
Price
$252.39
Log in pour la disponibilité
KIT(12items) Kit of 12
Product Details & Documents
Formalin Preserved
Grassfrog specimens and all dissection materials, including trays and pins, magnifiers, specimen storage bags, and dissection manual, are all included in one comprehensive package. Specimens are vacuum packed. The kit includes enough materials for 12 setups.
Spécifications
  • Désignation:
    WARD’S Grassfrog Dissection Lab
Accessories
Informations sur le produit
  • Catalog #
    Grade
    Size
    Availability
    Price
  • WARDSafe™
    WARDSafe™
    470233-546
    Laboratory
    19 L
    Log in pour la disponibilité
    Each (19l) - 345,85 $