Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Ward's® Slate (Red) | Avantor

Ward's® Slate (Red)

Slate (Red), bulk pack, 1 kg, Size will vary
Catalog # 470026-278
Supplier:   Avantor
Ward's® Slate (Red)
Catalog # 470026-278
Supplier:  Avantor
Ward's® Slate (Red)
Ward's® Slate (Red)

Slate (Red), bulk pack, 1 kg, Size will vary

Price
$31.49
Log in pour la disponibilité
Each
Product Details & Documents

Slaty cleavage; contains hematite.

  • Collected in New York
  • Multiple sizes and pack options to fit specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available

Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.

Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.

Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" - 2×3". Packaged in a labeled storage box for easy organization.

All Specimen dimensions are considered an average size range.

Spécifications
  • Désignation:
    Slate (Red), bulk pack, 1 kg
  • Specimen Dimensions:
    Size will vary