Quartzite
Quartzite, hand specimen, 2×2" - 3×4"
Catalog # 470026-176
Supplier:   Avantor
Quartzite
Catalog # 470026-176
Supplier:  Avantor
Quartzite
Quartzite
Quartzite, hand specimen, 2×2" - 3×4"
Price
$27.15
Log in pour la disponibilité
Each
Product Details & Documents
Fightly cemented, medium-to-coarse quartz grains.
  • Collected in South Dakotah
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Spécifications
  • Désignation:
    Quartzite, hand specimen
  • Specimen Dimensions:
    2×2" - 3×4"