Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Quartzite | Avantor

Quartzite

Quartzite, bulk pack; 1 kg, Size will vary
Catalog # 470026-174
Supplier:   Avantor
Quartzite
Catalog # 470026-174
Supplier:  Avantor
Quartzite
Quartzite

Quartzite, bulk pack; 1 kg, Size will vary

Price
$27.95
Log in pour la disponibilité
Each
Product Details & Documents

Fightly cemented, medium-to-coarse quartz grains.

  • Collected in South Dakotah
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available

Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.

Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.

Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.

All specimen dimensions are considered an average size range.

Spécifications
  • Désignation:
    Quartzite, bulk pack; 1 kg
  • Specimen Dimensions:
    Size will vary