Ward's® Obsidian
Obsidian, student specimens, 1×1 - 1×1"
Catalog # 470026-136
Supplier: Avantor
Jump to:
Product Details & Documents
Shiny black volcanic glass showing conchoidal fracture and some spherulites.
- Collected in Mexico
- Multiple sizes and pack options to fit your specific needs
- Cut by Ward's geologists to guarantee top market quality
- Matching thin section available
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Spécifications
- Désignation:Obsidian, student specimens
- Specimen Dimensions:1×1 - 1×1"