Ward's® Limestone (Shell)
Limestone (Shell), bulk pack, 1 kg, Size will vary
470026-082
: Avantor
Buff, coarse, crystalline “Austin Marble” with shell casts and molds.
Collected in Texas
- Multiple sizes and pack options to fit your specific needs
- Cut by Ward's geologists to guarantee top market quality
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
- Désignation:Limestone (Shell), bulk pack, 1 kg
- Specimen Dimensions:Size will vary