Ward's® Obsidian | Avantor
Ward's® Obsidian
Obsidian (thin section)
Catalog # 470024-984
Supplier:   Avantor
Ward's® Obsidian
Catalog # 470024-984
Supplier:  Avantor
Ward's® Obsidian
Ward's® Obsidian
Obsidian (thin section)
Price
$59.19
Log in pour la disponibilité
Each
Product Details & Documents
Shiny black volcanic glass showing conchoidal fracture and some spherulites.
  • Collected in Mexico
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Spécifications
  • Désignation:
    Obsidian (thin section)