Ward's® Metamorphic Petro-Lab Set | Avantor
Ward's® Metamorphic Petro-Lab Set
Metamorphic Rocks Petro-Lab Thin Sections Set, 7
Catalog # 470024-968
Supplier:   Avantor
Ward's® Metamorphic Petro-Lab Set
Catalog # 470024-968
Supplier:  Avantor
Ward's® Metamorphic Petro-Lab Set
Ward's® Metamorphic Petro-Lab Set
Metamorphic Rocks Petro-Lab Thin Sections Set, 7
Price
$657.99
Log in pour la disponibilité
Kit of 1
Product Details & Documents
Designed for Introductory College Petrography Labs.
  • Hand Made by Wards® staff Geologists
  • 7 Specially Selected Samples
The thin section set illustrates important mineral associations, textural features, and optical properties. Major mineralogical features are described in a worksheet that accompanies each slide. Using the basic information provided, students can determine optical properties, percentage estimations, and general compositional features that will assist them in their recognition of rocks and minerals. Matching hand specimens, average size 3×4", are available separately.
These seven slides cover low to high grade regional metamorphism. Among the key metamorphic indicator minerals present are actinolite, andalusite, chlorite, cordierite, garnet, graphite, kyanite, muscovite, sillimanite, and staurolite. The matching hand specimen set comes with numbered samples and an identification list. Each numbered sample comes in an individual plastic tray. Contents include: Schist, gneiss(×2), hornfels, phyllite, quartzite, staurolite.
Spécifications
  • Désignation:
    Metamorphic Rocks Petro-Lab Thin Sections Set
  • No. of Slides:
    7
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.