Ward's® Pleochroism Slide Thin Section | Avantor
Ward's® Pleochroism Slide Thin Section
Pleochroism slide thin section
Catalog # 470024-870
Ward's® Pleochroism Slide Thin Section
Catalog # 470024-870
Supplier:  
Ward's® Pleochroism Slide Thin Section
Ward's® Pleochroism Slide Thin Section
Pleochroism slide thin section
Discontinued
Product Details & Documents
Four mineral samples on one slide
  • A one of a kind slide
  • Created by Ward's geologists
  • Teach concepts that cannot be studied macroscopically
An innovative composite slide of four minerals displaying pleochroism; biotite, piedmontite, riebeckite, and tourmaline.
Ordering information: When minerals, rocks, and fossils cannot be accurately identified by macroscopic observation and testing, thin section slides can reveal the composition and structure of the specimens on a microscopic level. Thin section slides are precisely ground to a thickness of 0.03 mm across the entire specimen area. Mineral and rock thin sections are mounted on 27×46 mm microscope slides, and protected by a glass coverslip. All thin section slides are labeled for reference.
Spécifications
  • Désignation:
    Pleochroism slide thin section
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.