Ward's® Bulk Simulated Antisera | Avantor
Ward's® Bulk Simulated Antisera
WARD’S Bulk Simulated Antisera, Anti-A, 1 L
Catalog # 470024-646
Supplier:   Avantor
Ward's® Bulk Simulated Antisera
Catalog # 470024-646
Supplier:  Avantor
Ward's® Bulk Simulated Antisera
Ward's® Bulk Simulated Antisera
WARD’S Bulk Simulated Antisera, Anti-A, 1 L
Price
$71.10
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
At the request of our college customers, WARDS now offers these convenient bulk refills for our exclusive simulated ABO and Rh Blood-Typing Lab Activity (36 V 0019).
Instructions for performing blood typing using WARDS Simulated Blood and Simulated Antisera can be found in the teacher’s guide and student copymaster that accompanied the purchase of the lab activity. Each antiserum is provided in a one liter bottle.
Ordering information: This product is required to type WARD's simulated blood.
Spécifications
  • Désignation:
    WARD’S Bulk Simulated Antisera, Anti-A, 1 L