Ward's® ELISA Simulation Lab Activity | Avantor
Ward's® ELISA Simulation Lab Activity
ELISA simulation kit, 10
Catalog # 470002-904
Supplier:   Avantor
Ward's® ELISA Simulation Lab Activity
Catalog # 470002-904
Supplier:  Avantor
Ward's® ELISA Simulation Lab Activity
Ward's® ELISA Simulation Lab Activity
ELISA simulation kit, 10
Price
$197.95
Log in pour la disponibilité
Each Pallet of 36
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Safe and Colorful Activity
  • No refrigeration or expensive antibodies required
  • Teacher developed and classroom tested
  • Simulates the test for bacterial infection
  • Time Required: 45 minutes
  • Aligned to NGSS standards (HS: PS1, LS1)
Biology teacher, Brenda Royal, developed a quick and easy way to introduce concepts in immunology. Students perform a simulated ELISA test on fluid samples from two hypothetical patients to determine if either has been infected with an emerging bacterial strain. Materials for 10 student groups are included, along with teacher and student instructions.
Spécifications
  • Désignation:
    ELISA simulation kit
  • No. of Lab Groups:
    10