Preserved Turtles
Formalin Preserved Turtles, 6 to 8", Plain preserved, Pail of 10
  470000-582
 :  137.2126.10
Preserved Turtles
  470000-582
 :  
Preserved Turtles
Preserved Turtles
Formalin Preserved Turtles, 6 to 8", Plain preserved, Pail of 10
Formalin preserved turtles available in pails.
  • Reptile anatomy
  • General dissection
  • High quality preservation
We offer Pseudemys sp. turtles for study specimens that can be used to teach comparative anatomy and the similarities and differences between reptiles, amphibians, and mammals. Students can explore the structures of the shell, musculature, and internal organs of the study animal. The carapace is pre-cut for ease of dissection. We offer plain preserved, single, double, or triple injected turtles for your teaching or research needs.
  • Désignation:
    Formalin Preserved Turtles
  • Taille:
    6 to 8"
  • Injection Type:
    Plain preserved
  • Packaging:
    Pail of 10
Accessories
  •  
    Description
    Grade
    Size
  • 470233-546
    WARDSafe™
    Laboratory
    19 L
    Each (19l) - $345.85