Pipe Stem Triangles
Pipe Stem Triangle, side length 6 cm (2.5")
Catalog # 470189-156
Supplier: PRECISION MOULDED POLYMERS LIMITED
Supplier #: 1350627020000
Jump to:
Product Details & Documents
Triangles made from galvanized iron wire, with porcelain pipe stemming to reduce wire sagging and oxidation.
- Heavy gauge wire
- Galvanized steel
Provides a patform to hold items over a Bunsen burner.
Spécifications
- Désignation:Pipe Stem Triangle, side length 6 cm (2.5")