Pipe Stem Triangles
Pipe Stem Triangle, side length 4 cm (1.5")
  470189-154
 :  CPT015
Pipe Stem Triangles
  470189-154
Pipe Stem Triangles
Pipe Stem Triangles
Pipe Stem Triangle, side length 4 cm (1.5")
$4.90
Each Pack of 12
Triangles made from galvanized iron wire, with porcelain pipe stemming to reduce wire sagging and oxidation.
  • Heavy gauge wire
  • Galvanized steel
Provides a patform to hold items over a Bunsen burner.
  • Désignation:
    Pipe Stem Triangle, side length 4 cm (1.5")