Jars and Lids
Jar, Plastic, Wide-Mouth, 12 oz., (350 ml, 89 mm Diameter), Polystyrene, Clear
Catalog # 470188-600
Supplier: PLASTIC BOTTLES INC.
Supplier #: 192089R0000
Jump to:
Product Details & Documents
Stock up on classroom essentials for every classroom activity.
Spécifications
- Color:Clear
- Désignation:Jar, Plastic, Wide-Mouth, 12 oz., (350 ml, 89 mm Diameter)
- Matière:Polystyrene
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Screw Cap for Specimen Jar, 80 mm