$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
BAG PAPER 10X20X7.5CM (4X8X3) PK500
470188-584
:
REGIONAL DISTRIBUTORS, INC.
BAG PAPER 10X20X7.5CM (4X8X3) PK500
470188-584
:
REGIONAL DISTRIBUTORS, INC.
:
326
Frequently Bought Together
VWR® Disposable Serological Pipettes
: VWR International
Méthanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
VWR® Universal Pipette Tips
: VWR International