Jars and Lids | Avantor
Jars and Lids
Catalog # 470188-438
Jars and Lids
Catalog # 470188-438
Supplier:  
Jars and Lids
Jars and Lids
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
JAR POLYSTYRENE CLEAR 8OZ 89MM Catalog #470330-298
Product 470330-298 is a replacement for 470188-438 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Stock up on classroom essentials for every classroom activity.
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Screw Cap for Specimen Jar, 80 mm
    Screw Cap for Specimen Jar, 80 mm
    470175-872
    Log in pour la disponibilité
    Each - $2.95