Jars and Caps
70 mm, PET jar, 8 oz.
Catalog # 470184-336
Supplier: PLASTIC BOTTLES INC.
Fournisseur #: RM70WHT024
Jump to:
Product Details & Documents
Spécifications
- Capacité:8 oz.
- Désignation:70 mm, PET jar