Tooth Development Slides
Late dental gum slide, Section, Mallory Triple
Catalog # 470182-802
Supplier: Avantor
Jump to:
Product Details & Documents
Fetal pig tissue showing early tooth development.
- Identifying features clearly distinguishable
- Rigorous quality control standards
Slide samples created at various stages of mammal tooth-development.
Spécifications
- Désignation:Late dental gum slide
- Stain:Mallory Triple
- Orientation:Section