Ward's® Hay Medium Solution
Ward's® Hay Medium Solution
Hay Medium: Concentrated, Bottle, 125 mL
Catalog # 470180-766
Supplier:   Avantor
Ward's® Hay Medium Solution
Catalog # 470180-766
Supplier:  Avantor
CAS Number:  
Ward's® Hay Medium Solution
Hay Medium: Concentrated, Bottle, 125 mL
CAS Number:  
Price
$29.95
Log in for availability
Kit of 1
Product Details & Documents
Similar medium to Similar to Cereal Grass medium and Cerophyll. For culture of protists.
  • Sterile- no autoclaving
  • Convenient - prepared for you
  • Prepared consistenly every time
Concentrate comes with a vial of heat-treated wheat seeds.
Ordering information: These products contain living or perishable materials
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Specifications
Specifications
  • Type de conditionnement:
    Bottle
  • Désignation:
    Hay Medium: Concentrated
  • Taille:
    125 mL