Physarum Agar | Avantor
Physarum Agar
Prepared Physarum Agar Bottles
Catalog # 470180-730
Supplier:   MICROLOGY INC
Fournisseur #:  PA125
Physarum Agar
Catalog # 470180-730
Supplier:  MICROLOGY INC
CAS Number:  
Physarum Agar
Physarum Agar
Prepared Physarum Agar Bottles
CAS Number:  
Price
$63.10
Log in pour la disponibilité
Pack of 6
Product Details & Documents
With sterile oat flakes, for growth of Physarum polycephalum.
Spécifications
Spécifications
  • Désignation:
    Prepared Physarum Agar Bottles
Spécifications