Prepared Nutrient Agar
Nutrient Agar, Pour pack
Catalog # 470180-728
Supplier: Avantor
Jump to:
Product Details & Documents
General-purpose media support the growth of a variety of bacteria and fungi.
- Non-Selective, General-Purpose Media
- Available as Slant Tubes, Petri Plates, and Heat-and-Pour Bottles
- For Growing a Wide Range of Non-Fastidious Microorganisms
Nutrient agar, a non-selective media, is an excellent choice for growing a wide range of non-fastidious microorganisms. It is frequently used to isolate individual colonies from mixed cultures.
Tube media is configured as 12 tubes per pack. Petri dish media is configured as 10 plates per pack. Pour Paks come with three bottles of media (125 ml) and 20 sterile petri dishes. Just melt and pour.
Documents
Certificates
Recherche par numéro de lotSpécifications
Spécifications
- Type de conditionnement:Pour pack
- Désignation:Nutrient Agar