Crickets
Cricket nymphs, Pack of 12, ½"
  470180-336
 :   Avantor
Crickets
  470180-336
 :  Avantor
Crickets
Crickets
Cricket nymphs, Pack of 12, ½"
$18.90
Pack of 1
Cricket (Acheta domestica)or the Common House Cricket is available in adult and nymph sizes.
  • Exhibits incomplete metamorphosis
  • Excellent source of food for tarantulas and herps
 : These products contain living or perishable materials
 : Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
  • Désignation:
    Cricket nymphs, Pack of 12
  • Body Size:
    ½"