Ward's® Live Leeches
Live leeches, 1 - 2", Pack of 10
Catalog # 470180-296
 :  874721
Ward's® Live Leeches
Catalog # 470180-296
Ward's® Live Leeches
Ward's® Live Leeches
Live leeches, 1 - 2", Pack of 10
Price
$74.20
Log in pour la disponibilité
Pack of 10
Product Details & Documents
Erpobdella obscura
  • Freshwater leeches
Typical examples of the class Hirudinea, these freshwater leeches are dorsoventrally flattened with prominent terminal suckers.
The species supplied may be blood suckers, predators, or scavengers.
Ordering information: These products contain living or perishable materials. Seasonal item, available from April to October, depending on weather.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Spécifications
  • Désignation:
    Live leeches
  • Packaging:
    Pack of 10
  • Body Size:
    1 - 2"