Polystyrene Storage Jars
32 oz., 120 mm
Catalog # 470178-188
Supplier: Avantor
Jump to:
Product Details & Documents
Ideal for specimen storage.
- Clear polystyrene
- Wide-mouth
- Aligned with GLOBE protocols
Ordering information: Metal jar caps sold separately.
Specifications
- Taille du bouchon:120 mm
- Capacité:32 oz.