Ward's® Hay Medium Solution | Avantor
Ward's® Hay Medium Solution
Hay medium: Ready-to-use, Bottle, 1 Liter
Catalog # 470177-390
Supplier:   Avantor
Ward's® Hay Medium Solution
Catalog # 470177-390
Supplier:  Avantor
CAS Number:  
Ward's® Hay Medium Solution
Hay medium: Ready-to-use, Bottle, 1 Liter
CAS Number:  
Price
$62.93
Log in pour la disponibilité
Pack of 1
Product Details & Documents
Similar medium to Similar to Cereal Grass medium and Cerophyll. For culture of protists.
  • Sterile- no autoclaving
  • Convenient - prepared for you
  • Prepared consistenly every time
Concentrate comes with a vial of heat-treated wheat seeds.
Ordering information: These products contain living or perishable materials
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Spécifications
Spécifications
  • Type de conditionnement:
    Bottle
  • Désignation:
    Hay medium: Ready-to-use
  • Taille:
    1 Liter
Spécifications