Shell Vials with Corks
Shell vials, 1 dram, 15×45 mm, Oui, Cork cap
Catalog # 470177-984
Supplier: Avantor
Jump to:
Product Details & Documents
These multipurpose glass vials come with tightly fitting cork plugs.
Packaging: Package of 12.
Spécifications
- Avec bouchon:Oui
- Cap Type:Cork cap
- Désignation:Shell vials, 1 dram
- Diameter ×H:15×45 mm