Ward's® Introduction To Microscopes Slides
Ward's® Introduction To Microscopes Slides
Meat; raw and cooked, Sections, Hematoxylin and Eosin
Catalog # 470177-686
Supplier:   Avantor
Ward's® Introduction To Microscopes Slides
Catalog # 470177-686
Supplier:  Avantor
Ward's® Introduction To Microscopes Slides
Ward's® Introduction To Microscopes Slides
Meat; raw and cooked, Sections, Hematoxylin and Eosin
Price
$10.63
Log in for availability
Each
Product Details & Documents
Examine how a microscope works.
  • Professionally made microscope slides
  • Explore basic microscope functions and environmental materials
Introduce students to the world of microscopy and the world around them with these prepared slides. Use a selection of text slides to show students how microscopes change how you view materials. Students can then examine items that exist in the world around them.
Specifications
  • Désignation:
    Meat; raw and cooked
  • Stain:
    Hematoxylin and Eosin
  • Orientation:
    Sections