Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Ward's® Plasmid DNA for Transformations

pUC19 plus Loading Dye, 1 µg/100 µl
Catalog # 470177-184
Supplier:   Edvotek
 :  851306B
Ward's® Plasmid DNA for Transformations
Catalog # 470177-184
Supplier:  Edvotek
CAS Number:  
Ward's® Plasmid DNA for Transformations
Ward's® Plasmid DNA for Transformations

pUC19 plus Loading Dye, 1 µg/100 µl

CAS Number:  
Price
$44.15
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

These plasmids allow you to perform bacterial transformations, conferring new antibiotic resistance, as a visual way of demonstrating gene transfer.

  • Purchase only the amount of plasmid you need
  • Many plasmids already contain loading dye for added convenience

Plasmid pUC19: 2686 base pairs. Confers antibiotic resistance to ampicillin and complements defects in b-galactosidase in appropriate bacterial strains. Isolated from E. coli JM101.

pBR322: 4361 base pairs. Commonly used plasmid cloning vector for E. coli, and confers resistance to Ampicillin and Tetracycline.

 : These products are perishable materials, and are shipped next-day air on ice to ensure freshness. Please specify a delivery date at order time.

Specifications

Specifications

  • Concentration:
    1 µg/100 µl
  • Désignation:
    pUC19 plus Loading Dye

Specifications